Learn More
Sodium Hydroxide-Thiosulfate, 40%-2.5% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 40% (w/v) Sodium Hydroxide - 2.5% (w/v) Sodium Thiosulfate |
| Packaging | Natural Poly Bottle |
| pH | >13 |
| Quantity | 4 L |
| Chemical Name or Material | Sodium Hydroxide-Thiosulfate Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.