Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium hydroxymethanesulfinate dihydrate

Catalog No. 715301KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-530-1KG 1 kg
71-530-250G 250 g
71-530-5KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-530-1KG Supplier Sigma Aldrich Supplier No. 715301KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium hydroxymethanesulfinate dihydrate is a versatile reagent that can be used for a wide range of organic transformations such as:
  • A so2-2anion source for the preparation of sulfones and sultines
  • Debromination of vicinal dibromoalkanes
  • Reductive dehalogenation of aldehydes and ketones

Specifications

CAS 6035-47-8
Quantity 1 kg
Molecular Formula CH3NaO3S -+ 2H2O
Linear Formula CH3NaO3S -+ 2H2O
Synonym Rongalite; Sodium formaldehydesulfoxylate
Molecular Weight (g/mol) 154.12
Percent Purity ≥98.0% (RT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.