Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium β-hydroxypyruvate hydrate (Calc. based on dry substance, NT), ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111013975 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3975 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3975 Supplier MilliporeSigma Supelco Supplier No. 06367250MGF
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD16660324
Synonym beta-Hydroxypyruvic acid sodium salt; 3-Hydroxy-2-oxopropionic acid sodium salt

Specifications

CAS 3431-81-0
Linear Formula C3H3 NaO4 · xH2 O
Quantity 250 mg
Formula Weight 126.04 (anhydrous basis)
Percent Purity ≥97.0% (calc. based on dry substance, NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.