Learn More
GFS Chemicals Inc SODIUM HYPOCHLORITE SOLUTION, 5% AVAILABLE CHLORINE, REAGENT (ACS) | 500 ML | GFS CHEMICALS, INC. Item #: 67912 | Bulk #: 2763
SDP
Supplier:  GFS Chemicals Inc 67912
Encompass_Preferred
SODIUM HYPOCHLORITE SOLUTION, 5% AVAILABLE CHLORINE, REAGENT (ACS) | Chemical Formula : NaOCl | CAS # : 7681-52-9 | Specifications : Assay (available chlorine) (%) 5.0 % - Min, Sodium hydroxide (%) 2.5 %
Catalog No. NC9648304
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.