Learn More
LAB ALLEY Lab Alley Sodium Hypochlorite 5% Solution
SDP
Catalog No. NC2102985
Encompass
Sodium hypochlorite 5% Solution is an aqueous solution of Sodium Hypochlorite that may be used for cleaning disinfecting sanitizing household cleaning and the pressure/power washing of concrete pools and driveways. Household bleach is a solution containing 3–8% Sodium Hypochlorite by weight and 0.01–0.05% Sodium Hydroxide then the Sodium Hydroxide is used to slow the decomposition of Sodium Hypochlorite into Sodium Chloride and Sodium Chlorate.
This item is not returnable. View return policy
Catalog No. NC2102985 Supplier LAB ALLEY Supplier No. SHCN5500MLCS
Add to Cart
Add to Cart
Sodium hypochlorite 5% Solution is an aqueous solution of Sodium Hypochlorite that may be used for cleaning disinfecting sanitizing household cleaning and the pressure/power washing of concrete pools and driveways. Household bleach is a solution containing 3–8% Sodium Hypochlorite by weight and 0.01–0.05% Sodium Hydroxide then the Sodium Hydroxide is used to slow the decomposition of Sodium Hypochlorite into Sodium Chloride and Sodium Chlorate.