Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium iminodiacetate dibasic hydrate

Catalog No. 234877500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-487-7500G 500 g
23-487-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-487-7500G Supplier Sigma Aldrich Supplier No. 234877500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium iminodiacetate dibasic hydrate (iminodiacetic acid disodium salt monohydrate) has been used in the preparation of:
  • Different iminodiacetic acid-agarose supports, useful for selective adsorption of large proteins
  • N-(o-hydroxybenzyl) iminodiacetic acid (multidentante ligand)

Specifications

CAS 207398-95-6
Molecular Formula C4H5NNa2O4 -+ xH2O
Linear Formula C4H5NO4Na2 -+ xH2O
Quantity 500 g
MDL Number MFCD00150689
Synonym Iminodiacetic acid disodium salt hydrate
Molecular Weight (g/mol) 177.07 (anhydrous basis)
Percent Purity 98%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.