Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Iodate, Powder, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18613685 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-685 2.5 kg Amber Glass Bottle
18-613-682 25 g Amber Glass Bottle
18-613-679 100 g Amber Glass Bottle
18-613-680 125 g Amber Glass Bottle
18-613-683 500 g Amber Glass Bottle
18-613-681 12 kg Amber Glass Bottle
18-613-684 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-613-685 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S13252.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7681-55-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Percent Purity 99.5%
Grade Reagent
Loss on Drying 0.003
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Iodate, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.