Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraborate decahydrate, 99.5%, for analysis

Catalog No. AC205950100
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205950100 10 kg Plastic Drum
AC205950010 1 kg Plastic Bottle
AC205950050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205950100 Supplier Thermo Scientific Chemicals Supplier No. 205950100
Only null left
Add to Cart

CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
Molecular Weight (g/mol) 381.36
MDL Number MFCD00149193
Synonym Borax, Sodium borate decahydrate

Specifications

Melting Point 75.0°C
Color White
pH 9 to 9.5 (5% soln. at 20°C)
Physical Form Fine Crystalline Powder
Quantity 10 kg
Assay Percent Range 99.5%
Linear Formula Na2B4O7·10H2O
Fieser 04,461
Merck Index 15,8726
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 381.36
Percent Purity 99.5%
Grade Analytical
Packaging Plastic Drum
Chemical Name or Material Sodium tetraborate decahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility.
May damage the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.