Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Iodide AR Granular, Macron Fine Chemicals™

Catalog No. MK1141212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK1141-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK1141-212 Supplier Avantor Supplier No. 114106
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Specifications

CAS 7681-82-5
Physical Form Granules
Quantity 2.5 kg
Molecular Formula INa
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES [Na+].[I-]
IUPAC Name sodium;iodide
Molecular Weight (g/mol) 149.894
PubChem CID 5238
ChEBI CHEBI:33167
Formula Weight 149.89
Grade ACS
Packaging Glass
Chemical Name or Material Sodium Iodide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.