Learn More
Sodium iodide hydrate, Puratronic™, 99.999% (metals basis)
Description
Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline Aggregates |
| Quantity | 100 g |
| Solubility Information | Soluble in water (1840mg/ml at 25°C),alcohol,glycerol,and acetone. Insoluble in ether. |
| Molecular Weight (g/mol) | 149.89 |
| Formula Weight | 149.89 (Anhydrous) |
| Percent Purity | 99.999% |
| Grade | Puratronic |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Chemical Name or Material | Sodium iodide hydrate |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 231-679-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.