Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium iodoacetate, 305-53-3, MFCD00002686, 25 g
Catalog No. NC0916023
Encompass
Linear Formula: ICH2COONa, Molecular Weight: 207.93, ≥98%, Synonym: Iodoacetic acid sodium salt. Reagent for the modification of cysteine residues in proteins. Used in protein sequencing (including by Sanger in sequencing bovine insulin) to prevent oxidation of -SH sidechains. Reagent for the modification of cysteine residues in proteins. Used to induce animal osteoarthritis model system, which mimics both symptoms and histopathology of the human disease.
Catalog No. NC0916023 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I251225G
Linear Formula: ICH2COONa, Molecular Weight: 207.93, ≥98%, Synonym: Iodoacetic acid sodium salt. Reagent for the modification of cysteine residues in proteins. Used in protein sequencing (including by Sanger in sequencing bovine insulin) to prevent oxidation of -SH sidechains. Reagent for the modification of cysteine residues in proteins. Used to induce animal osteoarthritis model system, which mimics both symptoms and histopathology of the human disease.