Learn More
Medchemexpress LLC Sodium iodoacetate | 305-53-3 | MFCD00002686 | 99.9% | 1 G
SDP
Catalog No. 5000410437
Encompass_Preferred
Sodium iodoacetate is identified as a specific inhibitor of GAPDH, exhibiting glycolysis inhibitory activity. It is also known to induce osteoarthritis and related pain models in experimental animals.
  • Specific inhibitor of GAPDH.
  • Exhibits glycolysis inhibitory activity.
  • Induces osteoarthritis and related pain models in experimental animals.
Catalog No. 50-004-10437 Supplier Medchemexpress LLC Supplier No. HYD08491G
Sodium iodoacetate is identified as a specific inhibitor of GAPDH, exhibiting glycolysis inhibitory activity. It is also known to induce osteoarthritis and related pain models in experimental animals.
  • Specific inhibitor of GAPDH.
  • Exhibits glycolysis inhibitory activity.
  • Induces osteoarthritis and related pain models in experimental animals.