Learn More
Cayman Chemical SodIum Ionphore III 100mg
SDP
Catalog No. 502986954
Encompass_Preferred
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum
Catalog No. 50-298-6954 Supplier Cayman Chemical Supplier No. 22465100 MG
 Restricted Item
Add to Cart
Add to Cart
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum