Learn More
Cayman Chemical SodIum Ionphore III 10mg
SDP
Catalog No. 502987142
Encompass_Preferred
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum
Catalog No. 50-298-7142 Supplier Cayman Chemical Supplier No. 2246510 MG
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum