Learn More
Cayman Chemical Sodium Ionophore III, Purity: ≥98%, CAS: 81686-22-8, 50 mg
SDP
Catalog No. 502987558
Encompass_Preferred
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum
Catalog No. 50-298-7558 Supplier Cayman Chemical Supplier No. 2246550 MG
A sodium-selective ionophore; has been used as component of sodium-selective liquid membrane electrodes for the quantification of sodium in aqueous solutions and blood serum