Learn More
Cayman Chemical SodIum Ionphore VI 100mg
SDP
Catalog No. 502938266
Encompass_Preferred
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions
Catalog No. 50-293-8266 Supplier Cayman Chemical Supplier No. 27754100 MG
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions