Learn More
Cayman Chemical SodIum Ionphore VI 10mg
SDP
Catalog No. 502939522
Encompass_Preferred
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions
Catalog No. 50-293-9522 Supplier Cayman Chemical Supplier No. 2775410 MG
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions