Learn More
Cayman Chemical SodIum Ionphore VI 50mg
SDP
Catalog No. 502940966
Encompass_Preferred
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions
Catalog No. 50-294-0966 Supplier Cayman Chemical Supplier No. 2775450 MG
A sodium-selective ionophore that can be used in ion-selective electrodes to monitor sodium levels in aqueous solutions; selective for sodium over calcium, magnesium, lithium, ammonium, and potassium ions