Learn More
Medchemexpress LLC Sodium α-ketoisocaproate | 4502-00-5 | MFCD00064193 | 98.0% | 152.12 | C6H9NaO3 | 25 G

Supplier: Medchemexpress LLC 25GHYW017387
Sodium α-ketoisocaproate is the monosodium salt of α-ketoisocaproic acid, a leucine metabolite used as a biochemical reagent in metabolic and enzymatic studies. The compound is chemically defined with supporting analytical documentation and safety data available.
- High purity: 98.0% (assay by NMR)
- Chemical formula: C6H9NaO3
- Molar mass: 152.12 g/mol
- Commonly used as a leucine metabolite and metabolic reagent
- Supplied as a chemically defined sodium salt suitable for biochemical assays
- Certificate of analysis and safety data sheet available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.