Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium α-ketoisocaproate | 4502-00-5 | MFCD00064193 | 98.0% | 152.12 g/mol | C6H9NaO3 | 250 MG
SDP

Catalog No. 5000302733
Encompass_Preferred

Sodium α-ketoisocaproate is the sodium salt of α-ketoisocaproic acid, an endogenous metabolite of leucine used in biochemical and metabolic research. It is supplied for in vitro studies exploring metabolic pathways and insulin secretion mechanisms.

  • CAS number: 4502-00-5.
  • Molecular formula: C6H9NaO3.
  • Molecular weight: 152.12 g/mol.
  • Purity: 98.0%.
  • In vitro solubility: H2O 50 mg/mL (328.69 mM); may require ultrasonic assistance.
  • Storage: store at 4°C sealed and dry; in solvent -80°C (6 months) or -20°C (1 month) when sealed.
  • Typical use: metabolic studies, insulin secretion assays, and biochemical research.

Catalog No. 50-003-02733 Supplier Medchemexpress LLC Supplier No. 250MGHYW017387
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium α-ketoisocaproate is the sodium salt of α-ketoisocaproic acid, an endogenous metabolite of leucine used in biochemical and metabolic research. It is supplied for in vitro studies exploring metabolic pathways and insulin secretion mechanisms.

  • CAS number: 4502-00-5.
  • Molecular formula: C6H9NaO3.
  • Molecular weight: 152.12 g/mol.
  • Purity: 98.0%.
  • In vitro solubility: H2O 50 mg/mL (328.69 mM); may require ultrasonic assistance.
  • Storage: store at 4°C sealed and dry; in solvent -80°C (6 months) or -20°C (1 month) when sealed.
  • Typical use: metabolic studies, insulin secretion assays, and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.