Learn More
Medchemexpress LLC Sodium α-ketoisocaproic acid-13C2 | 2483736-09-8 | 98.0% | 154.11 | C413C2H9NaO3 | 1 MG
SDP
Catalog No. 5000299419
Encompass_Preferred
Sodium α-ketoisocaproic acid-13C2 is the 13C2 stable-isotope labeled sodium salt of α-ketoisocaproic acid intended for research use. It is used as a labeled tracer to study leucine metabolism and related biochemical pathways in metabolomics and analytical workflows such as mass spectrometry and NMR.
  • Provides 13C2 stable-isotope labeling for tracer studies.
  • Suitable for metabolomics, NMR, and MS-based assays.
  • High purity for reliable analytical results.
  • White to off-white solid, easy to handle and store.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-99419 Supplier Medchemexpress LLC Supplier No. HYW017387S11MG
Sodium α-ketoisocaproic acid-13C2 is the 13C2 stable-isotope labeled sodium salt of α-ketoisocaproic acid intended for research use. It is used as a labeled tracer to study leucine metabolism and related biochemical pathways in metabolomics and analytical workflows such as mass spectrometry and NMR.
  • Provides 13C2 stable-isotope labeling for tracer studies.
  • Suitable for metabolomics, NMR, and MS-based assays.
  • High purity for reliable analytical results.
  • White to off-white solid, easy to handle and store.
  • For research use only; not for human or clinical use.