Learn More
Medchemexpress LLC Sodium α-ketoisocaproic acid-13C6 | 2483736-03-2 | 99.3% | 158.08 | 13C6H9NaO3 | 1 MG
SDP
Catalog No. 5000300164
Encompass_Preferred
Sodium α-ketoisocaproic acid-13C6 is the uniformly 13C-labeled sodium salt of α-ketoisocaproic acid supplied as an analytical reference standard for tracer, metabolomics, and mass spectrometry or NMR studies. It is intended for use as an internal standard or labeled tracer to support quantitation and metabolic flux experiments.
  • Uniformly 13C6-labeled for clear isotopic tracing.
  • High purity (assay 99.3%) for accurate analytical results.
  • Suitable for MS and NMR applications as an internal or tracer standard.
  • Supplied as a neat solid enabling easy reconstitution for experiments.
  • Manufacturer provides SDS and COA for quality and regulatory use.
Catalog No. 50-003-00164 Supplier Medchemexpress LLC Supplier No. HYW017387S21MG
Sodium α-ketoisocaproic acid-13C6 is the uniformly 13C-labeled sodium salt of α-ketoisocaproic acid supplied as an analytical reference standard for tracer, metabolomics, and mass spectrometry or NMR studies. It is intended for use as an internal standard or labeled tracer to support quantitation and metabolic flux experiments.
  • Uniformly 13C6-labeled for clear isotopic tracing.
  • High purity (assay 99.3%) for accurate analytical results.
  • Suitable for MS and NMR applications as an internal or tracer standard.
  • Supplied as a neat solid enabling easy reconstitution for experiments.
  • Manufacturer provides SDS and COA for quality and regulatory use.