Learn More
Selleck Chemical LLC Sodium L-ascorbyl-2-phosphate S5115-25mg
SDP
Catalog No. 503110185
Encompass_Preferred
Sodium L-ascorbyl-2-phosphate (Sodium ascorbyl monophosphate Sodium ascorbyl phosphate SAP) is specifically produced for use as a stabilized source of vitamin C in cosmetic products It is used in skin care recipes for UV protection collagen production as an antioxidant and for its skin lightening and brightening effects Sodium L-ascorbyl-2-phosphate (2-Phospho-L-ascorbic acid trisodium salt L-Ascorbic acid 2-phosphate trisodium salt Sodium ascorbyl phosphate SAP) is a selective antioxidant and a stimulator of hepatocyte growth factor (HGF) production
Catalog No. 50-311-0185 Supplier Selleck Chemical LLC Supplier No. S511525MG
Sodium L-ascorbyl-2-phosphate (Sodium ascorbyl monophosphate Sodium ascorbyl phosphate SAP) is specifically produced for use as a stabilized source of vitamin C in cosmetic products It is used in skin care recipes for UV protection collagen production as an antioxidant and for its skin lightening and brightening effects Sodium L-ascorbyl-2-phosphate (2-Phospho-L-ascorbic acid trisodium salt L-Ascorbic acid 2-phosphate trisodium salt Sodium ascorbyl phosphate SAP) is a selective antioxidant and a stimulator of hepatocyte growth factor (HGF) production