Learn More
Eurofins Discoverx Sodium L-lactate, 10 mg. (92-1209)
Supplier:  Eurofins Discoverx 921209
Encompass
Sodium L-lactate is a sodium salt of lactic acid and an agonist for GPR81 (HCAR1). It is produced from pyruvate during anaerobic glycolysis and in proliferatively active cells as a result of reaction with the enzyme lactate dehydrogenase. Used as a food additive, it acts as a preservative, acidity regulator, and bulking agent. Also used in cleaning supplies like shampoo and soap, and can be used to treat arrhythmias, as a pressor sympathomimetic, and to prevent and control metabolic acidosis. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9735
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.