Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium L-lactate | 867-56-1 | MFCD00066576 | 98.0% | 112.06 g/mol | C3H5NaO3 | 5 MG
SDP

Catalog No. 5000208738
Encompass_Preferred

Sodium L-lactate (L-lactic acid sodium) is the sodium salt of L-lactic acid supplied as a solid analytical standard with ~98.0% purity. It is used as an analytical reference standard, a building block for polymer synthesis, and a reagent for research applications.

  • used as an analytical standard and reference material.
  • serves as a building block for polylactic acid and other syntheses.
  • white to off-white solid appearance.
  • purity about 98.0% and molecular weight 112.06 g/mol.
  • chemical formula C3H5NaO3, cas number 867-56-1.
  • available pack sizes typically 25 MG-500 MG; supplier-specific smaller packs may exist.

Catalog No. 50-002-08738 Supplier Medchemexpress LLC Supplier No. HYW040233R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium L-lactate (L-lactic acid sodium) is the sodium salt of L-lactic acid supplied as a solid analytical standard with ~98.0% purity. It is used as an analytical reference standard, a building block for polymer synthesis, and a reagent for research applications.

  • used as an analytical standard and reference material.
  • serves as a building block for polylactic acid and other syntheses.
  • white to off-white solid appearance.
  • purity about 98.0% and molecular weight 112.06 g/mol.
  • chemical formula C3H5NaO3, cas number 867-56-1.
  • available pack sizes typically 25 MG-500 MG; supplier-specific smaller packs may exist.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.