Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium L-Malate, MP Biomedicals

Catalog No. ICN218073 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218073 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218073 Supplier MP Biomedicals, Inc Supplier No. 0521807310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Formula C4H6Na2O5
Molecular Weight (g/mol) 180.067
InChI Key CBVOCNWSAAUNIY-JIZZDEOASA-N
Synonym l---malic acid disodium salt, l---malic acid disodium salt enzymatic
PubChem CID 124204296
IUPAC Name (2S)-2-hydroxybutanedioic acid;sodium
SMILES C(C(C(=O)O)O)C(=O)O.[Na].[Na]

Specifications

Quantity 10 g
Formula Weight 178.05
Chemical Name or Material Sodium L-Malate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.