Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lactate, 60% Syrup, BAKER™, J.T. Baker™

Catalog No. 02004165 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-165 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-165 Supplier Avantor Supplier No. V03408

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 72-17-3
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
InChI Key NGSFWBMYFKHRBD-UHFFFAOYSA-M
Synonym sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
PubChem CID 23666456
ChEBI CHEBI:75228
IUPAC Name sodium;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Na+]

Specifications

Quantity 500 mL
Formula Weight 112.06
Percent Purity 60%
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.31kg
Chemical Name or Material Sodium Lactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.