Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lactate Solution, FCC, 60%, Spectrum™ Chemical
SDP

Catalog No. 18606605 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-605 500 mL Amber Glass Bottle
18-606-606 4 L Amber Glass Bottle
18-606-607 20 L Poly Drum
18-606-608 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-605 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1217500ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1217, 72-17-3, C3H5NaO3

Spectrum™ Chemical Sodium Lactate Solution, 60 Percent, FCC is used in food products as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
InChI Key NGSFWBMYFKHRBD-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxypropanoate
SMILES [Na+].CC(O)C([O-])=O

Specifications

pH 5.0 to 9.0
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 58.8 to 61.2%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.