Learn More
Medchemexpress LLC Sodium laureth sulfate | 9004-82-4 | 70.3% | (C2H4O)nC12H26O4S.Na | 10 G
SDP
Catalog No. 5000432529
Encompass_Preferred
Sodium laureth sulfate is an anionic active agent with excellent properties for decontamination, emulsification, dispersion, wetting, and antifungal action. It is intended for research use only.
  • Anionic active agent
  • Possesses excellent decontamination properties
  • Possesses excellent emulsification properties
  • Possesses excellent dispersion properties
  • Possesses excellent wetting properties
  • Exhibits antifungal action
  • Enhances inactivation efficiency against S. cerevisiae cells
  • Disrupts membrane integrity and increases cell membrane permeability
  • Promotes ROS generation in S. cerevisiae cells
Catalog No. 50-004-32529 Supplier Medchemexpress LLC Supplier No. HYA027210G
Sodium laureth sulfate is an anionic active agent with excellent properties for decontamination, emulsification, dispersion, wetting, and antifungal action. It is intended for research use only.
  • Anionic active agent
  • Possesses excellent decontamination properties
  • Possesses excellent emulsification properties
  • Possesses excellent dispersion properties
  • Possesses excellent wetting properties
  • Exhibits antifungal action
  • Enhances inactivation efficiency against S. cerevisiae cells
  • Disrupts membrane integrity and increases cell membrane permeability
  • Promotes ROS generation in S. cerevisiae cells