Learn More
Medchemexpress LLC Sodium laureth sulfate | 9004-82-4 | 70.3% | (C2H4O)nC12H26O4S.Na | 50 G
SDP
Catalog No. 5000432230
Encompass_Preferred
Sodium laureth sulfate (70% in water) is an anionic active agent that possesses excellent decontamination, emulsification, dispersion, wetting, antifungal, and other properties.
  • Has excellent decontamination properties
  • Provides emulsification
  • Offers dispersion
  • Aids in wetting
  • Exhibits antifungal properties
Catalog No. 50-004-32230 Supplier Medchemexpress LLC Supplier No. HYA027250G
Sodium laureth sulfate (70% in water) is an anionic active agent that possesses excellent decontamination, emulsification, dispersion, wetting, antifungal, and other properties.
  • Has excellent decontamination properties
  • Provides emulsification
  • Offers dispersion
  • Aids in wetting
  • Exhibits antifungal properties