Learn More
Medchemexpress LLC Sodium laureth sulfate | 9004-82-4 | 70% | (C2H4O)nC12H26O4S.Na | 100 G
SDP
Catalog No. 5000432237
Encompass_Preferred
Sodium laureth sulfate (70% in water) is an anionic active agent known for its excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. This product is intended for research use only and is not sold to patients.
  • Possesses excellent decontamination properties
  • Offers emulsification and dispersion capabilities
  • Provides effective wetting and antifungal action
  • Enhances inactivation efficiency against S. cerevisiae cells when combined with plasma-activated water
  • Disrupts membrane integrity and increases cell membrane permeability in S. cerevisiae cells
  • Generates reactive oxygen species (ROS) in S. cerevisiae cells
  • Elevates activities of various lysosomal enzymes in animal models
Catalog No. 50-004-32237 Supplier Medchemexpress LLC Supplier No. HYA0272100G
Sodium laureth sulfate (70% in water) is an anionic active agent known for its excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. This product is intended for research use only and is not sold to patients.
  • Possesses excellent decontamination properties
  • Offers emulsification and dispersion capabilities
  • Provides effective wetting and antifungal action
  • Enhances inactivation efficiency against S. cerevisiae cells when combined with plasma-activated water
  • Disrupts membrane integrity and increases cell membrane permeability in S. cerevisiae cells
  • Generates reactive oxygen species (ROS) in S. cerevisiae cells
  • Elevates activities of various lysosomal enzymes in animal models