Learn More
Medchemexpress LLC Sodium laureth sulfate | 9004-82-4 | 70% | (C2H4O)nC12H26O4S.Na | 25 G
SDP
Catalog No. 5000358958
Encompass_Preferred
Sodium laureth sulfate (70% in water) is an anionic active agent that exhibits excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. It is intended for research use only.
  • Anionic active agent
  • Exhibits decontamination properties
  • Exhibits emulsification properties
  • Exhibits dispersion properties
  • Exhibits wetting properties
  • Provides antifungal properties
  • Enhances inactivation efficiency against *S. cerevisiae* cells
  • Disrupts membrane integrity in *S. cerevisiae* cells
  • Increases cell membrane permeability in *S. cerevisiae* cells
  • Elevates lysosomal enzyme activities in guinea pigs
Catalog No. 50-003-58958 Supplier Medchemexpress LLC Supplier No. 25GHYA0272
Add to Cart
Add to Cart
Sodium laureth sulfate (70% in water) is an anionic active agent that exhibits excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. It is intended for research use only.
  • Anionic active agent
  • Exhibits decontamination properties
  • Exhibits emulsification properties
  • Exhibits dispersion properties
  • Exhibits wetting properties
  • Provides antifungal properties
  • Enhances inactivation efficiency against *S. cerevisiae* cells
  • Disrupts membrane integrity in *S. cerevisiae* cells
  • Increases cell membrane permeability in *S. cerevisiae* cells
  • Elevates lysosomal enzyme activities in guinea pigs