Learn More
Medchemexpress LLC Sodium laureth sulfate | 9004-82-4 | 70% | (C2H4O)nC12H26O4S.Na | 5 G
SDP
Catalog No. 5000432889
Encompass_Preferred
Sodium laureth sulfate is an anionic active agent known for its excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. It is commonly used in various applications where these properties are beneficial.
  • Excellent decontamination and emulsification
  • Effective dispersion and wetting capabilities
  • Possesses antifungal properties
  • Suitable for a wide range of applications requiring anionic active agents
Catalog No. 50-004-32889 Supplier Medchemexpress LLC Supplier No. HYA02725G
Sodium laureth sulfate is an anionic active agent known for its excellent decontamination, emulsification, dispersion, wetting, and antifungal properties. It is commonly used in various applications where these properties are beneficial.
  • Excellent decontamination and emulsification
  • Effective dispersion and wetting capabilities
  • Possesses antifungal properties
  • Suitable for a wide range of applications requiring anionic active agents