Learn More
Medchemexpress LLC Sodium lauroyl glutamate | 29923-31-7 | 95.8% | 1 G
SDP
Catalog No. 5000338364
Encompass_Preferred
Sodium lauroyl glutamate is an anionic amino acid surfactant used primarily in cleaning products. It is considered a mild surfactant with low irritation to nude mouse skin, exhibiting a pH value of 5.41. It also has potential for irritant contact dermatitis and possible anti-irritating effects in surfactant mixtures on human skin.
  • Anionic amino acid surfactant
  • Mild surfactant properties
  • Ideal for cleaning product formulations
  • Low irritation potential
  • pH value of 5.41
Catalog No. 50-003-38364 Supplier Medchemexpress LLC Supplier No. HY1303131G
Sodium lauroyl glutamate is an anionic amino acid surfactant used primarily in cleaning products. It is considered a mild surfactant with low irritation to nude mouse skin, exhibiting a pH value of 5.41. It also has potential for irritant contact dermatitis and possible anti-irritating effects in surfactant mixtures on human skin.
  • Anionic amino acid surfactant
  • Mild surfactant properties
  • Ideal for cleaning product formulations
  • Low irritation potential
  • pH value of 5.41