Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzaldehyde, ACS reagent

Catalog No. AC423630250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423630250 25 g Glass bottle
AC423631000 100 g Glass bottle
AC423635000 500 g Glass bottle
AC423630025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423630250 Supplier Thermo Scientific Chemicals Supplier No. 423630250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100-10-7 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Color White to Off-white
Density 5.15
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 25 g
Beilstein 14, 31
Fieser 01,273; 02,146
Merck Index 15, 3252
Solubility Information Solubility in water: 0.3g/L (20°C) Other solubilities: Soluble in acetic acid, Soluble in alcohol, ether and chloroform
Formula Weight 149.19
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less
Hazard Category Signal Word: WARNING!
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
EINECSNumber 202-819-0
RTECSNumber CU5775000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.