Learn More
U.S. Pharmacopeia Sodium lauryl sulfate, 151-21-3, MFCD00036175, 1 g
Catalog No. NC1411969
Encompass
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, mp: 204-207 °C, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium dodecyl sulfate, Sodium lauryl sulfate.
Catalog No. NC1411969 Supplier U.S. Pharmacopeia Supplier No. 1614363
Linear Formula: CH3(CH2)11OSO3Na, Molecular Weight: 288.38, mp: 204-207 °C, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium dodecyl sulfate, Sodium lauryl sulfate.