Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lauryl Sulfate, 20% (w/v) Solution, Spectrum™ Chemical
SDP

Catalog No. 18606610 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-610 20 L Poly Pail
18-606-609 500 mL Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-610 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S134320LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1343, 151-21-3, C12H25O4SNa

Spectrum™ Chemical Sodium Lauryl Sulfate, 20 Percent (w/v) Solution , is an aqueous solution of an organic compound, and anionic surfactant derived from coconut or palm oils, for use as a detergent. As an Organosulfate, this salt has the amphiphilic properties needed in a detergent. It's uses vary from shark repellent, fruit fly killer and biocide.

Chemical Identifiers

CAS 151-21-3 , 7732-18-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

CAS Max % 0.2, 0.8
Concentration 20%
Quantity 20 L
Packaging Poly Pail
Grade Ungraded
Chemical Name or Material Sodium Lauryl Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.