Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Sodium Lauryl Sulfate NF
SDP

Catalog No. 14185630 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-630 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-630 Supplier Medisca Supplier No. 062208
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Lauryl Sulfate NF comes as small, white or light yellow crystals. Has a slight, characteristic odor.

Sodium Lauryl Sulfate NF comes as small, white or light yellow crystals. Has a slight, characteristic odor.

Alternative name(s): Sodium Dodecyl Sulfate, sodium dodecylsulfate, sodium lauryl sulphate,sodium dodecyl sulphate

Specifications

Chemical Name or Material Sodium Lauryl Sulfate NF
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 151-21-3
Type Drug Standard
Molecular Weight (g/mol) 288.4
Quantity 500 g
Percent Purity ≥ 85.0 %
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.