Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lauryl Sulfate, NF, EP, BP, JP, Spectrum™ Chemical
SDP

Catalog No. 18606620 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-620 12 kg Fiber Drum
18-606-621 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-620 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO18012KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

SO180, 151-21-3, C12H25O4SNa

Spectrum™ Chemical Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 12 kg
Packaging Fiber Drum
Water 0.05
Percent Purity 85%
Grade Multi-Compendial
Chemical Name or Material Sodium Lauryl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.