Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lauryl Sulfate, Reagent, 97%, Spectrum™ Chemical
SDP

Catalog No. 18606623 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-623 100 g Amber Glass Bottle
18-606-624 500 g Amber Glass Bottle
18-606-625 1 kg Amber Glass Bottle
18-606-626 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-623 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1333100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1333, 151-21-3, C12H25O4SNa

Spectrum™ Chemical Sodium Lauryl Sulfate, Reagent , is an organic compound, and anionic surfactant derived from coconut or palm oils, for use as a detergent. As an Organosulfate, this salt has the amphiphilic properties needed in a detergent. It's uses vary from shark repellent, fruit fly killer and biocide. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 100 g
Packaging Amber Glass Bottle
Water 0.05
Percent Purity 97%
Grade Reagent
Chemical Name or Material Sodium Lauryl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.