Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Lauryl Sulfate (SLS), 10% Solution, Innovating Science™
SCI_ED

Catalog No. S000045 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000045 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000045 Supplier Aldon Corporation Supplier No. IS28144
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Sodium lauryl sulfate
CAS 7732-18-5, 151-21-3
Physical Form Pellets
Quantity 100 mL
Grade Reagent
Concentration or Composition (by Analyte or Components) 10%
Packaging HDPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.