Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium malonate | 141-95-7 | MFCD00002708 | 98.0% | 148.03 g/mol | C3H2Na2O4 | 25 G
SDP

Catalog No. 5000281876
Encompass_Preferred

Sodium malonate is a research-grade organic salt used as an intermediate and reagent in the synthesis of active compounds and small-molecule research. It is supplied as a white to off-white solid with documented purity and handling instructions suitable for laboratory use.

  • Used as a drug intermediate for organic synthesis.
  • White to off-white solid form.
  • Purity 98.0%.
  • Molecular weight 148.03 g/mol.
  • Chemical formula C3H2Na2O4.
  • CAS number 141-95-7.
  • Store sealed at 4°C, away from moisture; solution storage -80°C (6 months) or -20°C (1 month).
  • For research use only.

Catalog No. 50-002-81876 Supplier Medchemexpress LLC Supplier No. HYW28372625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium malonate is a research-grade organic salt used as an intermediate and reagent in the synthesis of active compounds and small-molecule research. It is supplied as a white to off-white solid with documented purity and handling instructions suitable for laboratory use.

  • Used as a drug intermediate for organic synthesis.
  • White to off-white solid form.
  • Purity 98.0%.
  • Molecular weight 148.03 g/mol.
  • Chemical formula C3H2Na2O4.
  • CAS number 141-95-7.
  • Store sealed at 4°C, away from moisture; solution storage -80°C (6 months) or -20°C (1 month).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.