Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium malonate | 141-95-7 | MFCD00002708 | 98.0% | 148.03 g·mol⁻¹ | C3H2Na2O4 | 50 G
SDP

Catalog No. 5000280539
Encompass_Preferred

Sodium malonate is the disodium salt of malonic acid used as an intermediate in organic synthesis and for laboratory research. It is supplied as a white to off-white solid with high purity suitable for a range of chemical synthesis applications.

  • High purity: 98.0%.
  • White to off-white solid appearance.
  • Molecular formula C3H2Na2O4.
  • Molecular weight 148.03 g·mol⁻¹.
  • CAS number 141-95-7.
  • Available in laboratory-scale quantities such as 50 G.

Catalog No. 50-002-80539 Supplier Medchemexpress LLC Supplier No. HYW28372650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium malonate is the disodium salt of malonic acid used as an intermediate in organic synthesis and for laboratory research. It is supplied as a white to off-white solid with high purity suitable for a range of chemical synthesis applications.

  • High purity: 98.0%.
  • White to off-white solid appearance.
  • Molecular formula C3H2Na2O4.
  • Molecular weight 148.03 g·mol⁻¹.
  • CAS number 141-95-7.
  • Available in laboratory-scale quantities such as 50 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.