Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium malonate dibasic

Catalog No. 63409500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-409-500G 500 g
63-409-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-409-500G Supplier Sigma Aldrich Supplier No. 63409500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium malonate dibasic can be use in the synthesis of sodium carbonate and sodium oxalate via wet oxidation and catalytic wet oxidation
  • It can also be used to synthesize malonato-bridged manganese(ii)complexes like [mn(mal)(h2o)2]n and [mn2(mal)2(4,4'-bipy)(h2o)2]n,(bipy =bipyridine, mal =malonate), which have the tendency to show weak anti-ferromagnetic effect

Specifications

CAS 141-95-7
Molecular Formula C3H2Na2O4
Linear Formula CH2(COONa)2
MDL Number MFCD00002708
Quantity 500 g
Synonym Disodium malonate; Malonic acid disodium salt
Molecular Weight (g/mol) 148.03
Percent Purity ≥97.0% (NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.