Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium malonate dibasic monohydrate

Catalog No. M187525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M187525G 25 g
M1875100G 100 g
M1875500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M187525G Supplier Sigma Aldrich Supplier No. M187525G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium malonate dibasic monohydrate

Specifications

CAS 26522-85-0
Molecular Formula C3H2Na2O4 -+ H2O
Linear Formula CH2(CO2Na)2 -+ H2O
MDL Number MFCD00002708
Quantity 25 g
Synonym Malonic acid disodium salt monohydrate; Propanedioic acid disodium salt monohydrate
Molecular Weight (g/mol) 166.04
Percent Purity ≥98% (non-aqueous titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.