Learn More
Medchemexpress LLC Sodium malonate hydrate | 26522-85-0 | MFCD00002708 | 99.1% | 166.04 g·mol-1 | C3H4Na2O5 | 100 G
SDP
Catalog No. 5000300925
Encompass_Preferred
Sodium malonate hydrate is the disodium salt of malonic acid supplied as a research-grade, crystalline reagent suitable for biochemical and synthetic applications. Its defined hydration state and high assay purity support reproducible results in buffer formulations, metal-chelation assays, and as a building block in organic synthesis.
  • High assay purity (99.07%).
  • Molecular formula C3H4Na2O5; molecular weight 166.04 g·mol-1.
  • White crystalline solid with good water solubility.
  • Useful as a buffer component and building block in organic synthesis.
  • Functions as a chelator for metal ions in biochemical assays.
  • Available in multiple pack sizes for small- and bulk-scale needs.
Catalog No. 50-003-00925 Supplier Medchemexpress LLC Supplier No. HYW017165100G
Sodium malonate hydrate is the disodium salt of malonic acid supplied as a research-grade, crystalline reagent suitable for biochemical and synthetic applications. Its defined hydration state and high assay purity support reproducible results in buffer formulations, metal-chelation assays, and as a building block in organic synthesis.
  • High assay purity (99.07%).
  • Molecular formula C3H4Na2O5; molecular weight 166.04 g·mol-1.
  • White crystalline solid with good water solubility.
  • Useful as a buffer component and building block in organic synthesis.
  • Functions as a chelator for metal ions in biochemical assays.
  • Available in multiple pack sizes for small- and bulk-scale needs.