Learn More
Medchemexpress LLC Sodium malonate hydrate | 26522-85-0 | MFCD00002708 | 99.1% | 166.04 g/mol | C3H4Na2O5 | 50 G
SDP
Catalog No. 5000300152
Encompass_Preferred
Sodium malonate hydrate is the disodium salt hydrate of malonic acid used as a reagent and metal-chelating agent in analytical chemistry and life-science research. It is supplied as a crystalline hydrate with batch-specific documentation (COA and SDS), and is intended for laboratory research use.
  • Complexing agent for determination of beryllium and copper.
  • High assay purity (≈99.1%).
  • Monohydrate form; molecular formula C3H4Na2O5; molecular weight 166.04 g/mol.
  • Supplied in multiple pack sizes suitable for laboratory use.
  • COA and SDS available for batch-specific documentation.
Catalog No. 50-003-00152 Supplier Medchemexpress LLC Supplier No. HYW01716550G
Sodium malonate hydrate is the disodium salt hydrate of malonic acid used as a reagent and metal-chelating agent in analytical chemistry and life-science research. It is supplied as a crystalline hydrate with batch-specific documentation (COA and SDS), and is intended for laboratory research use.
  • Complexing agent for determination of beryllium and copper.
  • High assay purity (≈99.1%).
  • Monohydrate form; molecular formula C3H4Na2O5; molecular weight 166.04 g/mol.
  • Supplied in multiple pack sizes suitable for laboratory use.
  • COA and SDS available for batch-specific documentation.