Learn More
Medchemexpress LLC Sodium malonate hydrate | 26522-85-0 | MFCD00002708 | 99.1% | 166.04 g/mol | C3H4Na2O5 | 500 G
SDP
Catalog No. 5000300554
Encompass_Preferred
Sodium malonate hydrate is the disodium salt monohydrate of malonic acid used as a complexing agent in analytical chemistry and as a reagent in life-science research. It is supplied as a high-purity, easy-to-handle solid suitable for laboratory use.
  • High purity (99.07%) for reliable analytical results.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Molecular weight 166.04 g/mol; formula C3H4Na2O5 for accurate stoichiometry.
  • Useful as a complexing agent for the determination of beryllium and copper.
  • Available in 500 g packaging for bulk laboratory applications.
Catalog No. 50-003-00554 Supplier Medchemexpress LLC Supplier No. HYW017165500G
Sodium malonate hydrate is the disodium salt monohydrate of malonic acid used as a complexing agent in analytical chemistry and as a reagent in life-science research. It is supplied as a high-purity, easy-to-handle solid suitable for laboratory use.
  • High purity (99.07%) for reliable analytical results.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Molecular weight 166.04 g/mol; formula C3H4Na2O5 for accurate stoichiometry.
  • Useful as a complexing agent for the determination of beryllium and copper.
  • Available in 500 g packaging for bulk laboratory applications.