Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium (meta)periodate

Catalog No. 769517100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-951-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-951-7100G Supplier Sigma Aldrich Supplier No. 769517100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful reagent for the oxidation of carbohydrates prior to labeling with biotin hydrazide

Specifications

CAS 7790-28-5
Molecular Formula INaO4
Linear Formula NaIO4
Quantity 100 g
MDL Number MFCD00003534
Synonym Sodium periodate
Molecular Weight (g/mol) 213.89
Percent Purity ≥99%; 99.0-101.0% (RT)
Biological Activity thymus gland
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.