Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium (Meta)Periodate, Solstice

Catalog No. 6001371 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-71 500 g
60-045-816 6 x 500 g
60-013-70 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-013-71 Supplier Solstice Supplier No. 30323500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Quantity 500 g
Linear Formula NaIO4
UN Number 3085
Formula Weight 213.89g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium (Meta)Periodate, Reag. Ph. Eur. reag. ISO
Hazard Statement P220-P273-P280-P305 + P351 + P338-P310
Precautionary Statement H271-H314-H372-H400
EINECSNumber 232-197-6
RTECSNumber SD4550000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.